3-Bromo-N-methylbenzamide 98%

Supplier: Ambeed
Warning

A537072-100MG A537072-10G A537072-1G A537072-250MG A537072-5G A537072-25G
77034-784EA 5 USD
77034-784 77034-786 77034-788 77034-790 77034-792 77872-899
3-Bromo-N-methylbenzamide 98%
3-Bromo-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MDL Number: MFCD01060879
CAS Number: 49834-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR