2-(1H-Benzo[d]imidazol-2-yl)acetic acid 95%

Supplier: Ambeed
Warning

A537069-10G A537069-1G A537069-250MG A537069-5G
77017-542EA 302 USD
77017-542 77017-544 77017-546 77017-548
2-(1H-Benzo[d]imidazol-2-yl)acetic acid 95%
2-(1H-Benzo[d]imidazol-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD01102656
CAS Number: 13570-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR