1-Bromo-4-chloro-2-fluoro-5-methylbenzene 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-chloro-4-fluorotoluene

A536990-100G A536990-10G A536990-25G
76991-568EA 217.49 USD
76991-568 76991-570 76991-572
1-Bromo-4-chloro-2-fluoro-5-methylbenzene 95%
1-Bromo-4-chloro-2-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClF
MW: 223.47 g/mol
MDL Number: MFCD09800685
CAS Number: 200190-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR