4-(4'-Methoxybenzyloxy)phenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (4-((4-Methoxybenzyl)oxy)phenyl)boronic acid

A536697-1G A536697-5G
76991-564EA 38.2 USD
76991-564 76991-566
4-(4'-Methoxybenzyloxy)phenylboronic acid 96%
4-(4'-Methoxybenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BO₄
MW: 258.0800 g/mol
MDL Number: MFCD09038417
CAS Number: 156635-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR