tert-Butyl-N-{3-[(tert-butoxycarbonyl)amino]-2-hydroxypropyl}carbamate 95%

Supplier: Ambeed
Warning

A536051-100MG A536051-250MG
77004-982EA 22 USD
77004-982 77004-984
tert-Butyl-N-{3-[(tert-butoxycarbonyl)amino]-2-hydroxypropyl}carbamate 95%
tert-Butyl-N-{3-[(tert-butoxycarbonyl)amino]-2-hydroxypropyl}carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₆N₂O₅
MDL Number: MFCD12498706
CAS Number: 98642-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR