(3-Bromo-2-butoxy-5-fluorophenyl)boronic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-2-butoxy-5-fluorophenylboronic acid
A535806-1G
A535806-250MG
A535806-5G
77017-528EA
83
USD
77017-528
77017-530
77017-532
(3-Bromo-2-butoxy-5-fluorophenyl)boronic acid 95%
(3-Bromo-2-butoxy-5-fluorophenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BBrFO₃ |
MDL Number:
MFCD06411282 CAS Number: 352534-85-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...