1-Methyluracil 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methylpyrimidine-2,4(1H,3H)-dione

A535440-100MG A535440-1G A535440-250MG A535440-5G
77017-522EA 36.08 USD
77017-522 77017-524 77017-526 77569-036
1-Methyluracil 98%
1-Methyluracil

Precaución: For research use only.

Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
MDL Number: MFCD00038666
CAS Number: 615-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR