3-Iodo-4-methyl-1H-indazole 97%

Supplier: Ambeed
Warning

A534415-250MG A534415-5G
77034-780EA 33.95 USD
77034-780 77034-782
3-Iodo-4-methyl-1H-indazole 97%
3-Iodo-4-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇IN₂
MW: 258.0591 g/mol
MDL Number: MFCD07781537
CAS Number: 885522-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR