4-Ethyl-4-methylmorpholinium bromide 97%

Supplier: Ambeed
Danger

A534261-5G A534261-25G
76991-546EA 18 USD
76991-546 77594-650
4-Ethyl-4-methylmorpholinium bromide 97%
4-Ethyl-4-methylmorpholinium bromide

Precaución: for Research Use Only

Formula: C₇H₁₆BrNO
MDL Number: MFCD03095385
CAS Number: 65756-41-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR