(4-Methylbenzyl)hydrazine dihydrochloride 95%

Supplier: Ambeed
Warning

A533987-100MG A533987-250MG
77004-968EA 12 USD
77004-968 77004-970
(4-Methylbenzyl)hydrazine dihydrochloride 95%
(4-Methylbenzyl)hydrazine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₄Cl₂N₂
MW: 209.12 g/mol
MDL Number: MFCD06800973
CAS Number: 1085453-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR