4-Bromoisoquinoline-1-carbonitrile 95%

Supplier: Ambeed
Warning

A533306-100MG A533306-250MG A533306-1G
77017-502EA 15 USD
77017-502 77017-504 77891-078
4-Bromoisoquinoline-1-carbonitrile 95%
4-Bromoisoquinoline-1-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₅BrN₂
MDL Number: MFCD11506148
CAS Number: 27224-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR