5-Amino-2-(4-methylpiperazin-1-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A532820-100MG A532820-1G A532820-250MG A532820-5G
77004-960EA 106.09 USD
77004-960 77004-962 77004-964 77004-966
5-Amino-2-(4-methylpiperazin-1-yl)benzonitrile 98%
5-Amino-2-(4-methylpiperazin-1-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₄
MW: 216.28 g/mol
MDL Number: MFCD11852581
CAS Number: 288251-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR