1-Naphthyl boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Naphthalene-1-boronic acid Pinacol ester, 1-Naphthalenboronic acid, pinacol ester, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene, 4,4,5,5-Tetramethyl-2-(1-naphthyl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborolane

A531829-100G A531829-1G A531829-250MG A531829-25G A531829-5G
77034-764EA 148 USD
77034-764 77034-766 77034-768 77034-770 77034-772
1-Naphthyl boronic acid pinacol ester 98%
1-Naphthyl boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₁₉BO₂
MW: 254.1319 g/mol
MDL Number: MFCD05663869
CAS Number: 68716-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR