4-Bromo-7-(trifluoromethyl)-1H-indazole 95%

Supplier: Ambeed
Warning

A531557-100MG A531557-1G A531557-250MG
77017-492EA 78.51 USD
77017-492 77017-494 77017-496
4-Bromo-7-(trifluoromethyl)-1H-indazole 95%
4-Bromo-7-(trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
MDL Number: MFCD16658657
CAS Number: 1186334-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR