1-(4-methylidenepiperidino)ethan-1-one 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Methylenepiperidin-1-yl)ethanone

A531225-100MG A531225-1G A531225-250MG
77017-486EA 85.94 USD
77017-486 77017-488 77017-490
1-(4-methylidenepiperidino)ethan-1-one 95%
1-(4-methylidenepiperidino)ethan-1-one

Precaución: for Research Use Only

Formula: C₈H₁₃NO
MW: 139.195 g/mol
MDL Number: MFCD00831797
CAS Number: 308087-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR