MLN-8054 98+%

Supplier: Ambeed
Warning

Synonyms: MLN8054, 4-[[9-chloro-7-(2,6-difluorophenyl)-5H-pyrimido[5,4-d][2]benzazepin-2-yl]amino]benzoic acid, 4-((9-Chloro-7-(2,6-difluorophenyl)-5H-benzo[c]pyrimido[4,5-e]azepin-2-yl)amino)benzoic acid

A531108-50MG A531108-100MG A531108-250MG
77034-756EA 141 USD
77034-756 77602-270 77669-358
MLN-8054 98+%
MLN-8054

Precaución: for Research Use Only

Formula: C₂₅H₁₅ClF₂N₄O₂
MDL Number: MFCD11840561
CAS Number: 869363-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR