1-Bromo-2-fluoro-4-(methylsulfonyl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-fluorophenyl methyl sulphone

A530897-100MG A530897-1G A530897-5G
77034-754EA 15.92 USD
77034-754 77602-268 77673-414
1-Bromo-2-fluoro-4-(methylsulfonyl)benzene 97%
1-Bromo-2-fluoro-4-(methylsulphonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₆BrFO₂S
MW: 253.0887 g/mol
MDL Number: MFCD11053812
CAS Number: 1032825-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR