3,5-Bis(difluoromethyl)-1H-pyrazole 98%
Supplier: Ambeed
|
Danger
|
A530746-1G
A530746-250MG
77004-940EA
230.22
USD
77004-940
77602-266
3,5-Bis(difluoromethyl)-1H-pyrazole 98%
3,5-Bis(difluoromethyl)-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₅H₄F₄N₂ |
MDL Number:
MFCD03419821 CAS Number: 77614-79-0 UN: 1759 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...