3-(1H-Pyrazol-1-yl)propan-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: 3-(1-Pyrazolyl)-1-propanol

A530618-1G A530618-250MG A530618-25G A530618-5G
77004-932EA 46.68 USD
77004-932 77004-934 77004-936 77004-938
3-(1H-Pyrazol-1-yl)propan-1-ol 98%
3-(1H-Pyrazol-1-yl)propan-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O
MW: 126.16 g/mol
MDL Number: MFCD03727229
CAS Number: 180741-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR