Ethyl 3-Cyclohexyl-3-oxopropionate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-Cyclohexyl-3-oxopropanoate, 3-Cyclohexyl-3-oxopropionic acid Ethyl ester

A530584-1G A530584-250MG A530584-5G
77004-928EA 30.77 USD
77004-928 77004-930 77602-264
Ethyl 3-Cyclohexyl-3-oxopropionate 95%
Ethyl 3-Cyclohexyl-3-oxopropionate

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₃
MW: 198.26 g/mol
MDL Number: MFCD04115574
CAS Number: 15971-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR