3-Chloro-4-(2-chlorophenoxy)aniline 97%

Supplier: Ambeed
Warning

A530389-5G A530389-1G A530389-250MG
77004-926EA 144 USD
77004-926 77673-412 77882-295
3-Chloro-4-(2-chlorophenoxy)aniline 97%
3-Chloro-4-(2-chlorophenoxy)aniline

Precaución: for Research Use Only

Formula: C₁₂H₉Cl₂NO
MDL Number: MFCD07365123
CAS Number: 56966-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR