N-(Furan-2-ylmethyl)ethanamine 95%

Supplier: Ambeed
Warning

Synonyms: N-(2-Furylmethyl)ethanamine

A530222-100MG A530222-1G
77034-750EA 58.35 USD
77034-750 77034-752
N-(Furan-2-ylmethyl)ethanamine 95%
N-(Furan-2-ylmethyl)ethanamine

Precaución: for Research Use Only

Formula: C₇H₁₁NO
MW: 125.17 g/mol
MDL Number: MFCD00069030
CAS Number: 14496-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR