Bis(p-tolyl)phosphine oxide 98%

Supplier: Ambeed
Warning

Synonyms: Di-p-tolylphosphine oxide, Di(p-tolyl)phosphine Oxide

A530217-10G A530217-1G A530217-25G A530217-5G A530217-100G
77034-742EA 146.41 USD
77034-742 77034-744 77034-746 77034-748 77669-352
Bis(p-tolyl)phosphine oxide 98%
Bis(p-tolyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₁₄H₁₅OP
MW: 230.24 g/mol
MDL Number: MFCD01445489
CAS Number: 2409-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR