3-Amino-4-hydroxyphenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-hydroxyphenylboronic acid, pinacol ester, 2-Amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

A529957-1G A529957-250MG A529957-5G
77017-472EA 185.66 USD
77017-472 77017-474 77017-476
3-Amino-4-hydroxyphenylboronic acid pinacol ester 95%
3-Amino-4-hydroxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₃
MW: 235.09 g/mol
MDL Number: MFCD13176535
CAS Number: 760990-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR