7-Bromopyrazolo[1,5-a]pyridine 96%

Supplier: Ambeed
Warning

A529830-100MG A529830-1G A529830-5G A529830-250MG
77017-466EA 80.63 USD
77017-466 77017-468 77017-470 77613-660
7-Bromopyrazolo[1,5-a]pyridine 96%
7-Bromopyrazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
MDL Number: MFCD06739220
CAS Number: 885275-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR