6-Chloro-3-hydroxybenzo[b]thiophene-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A529682-100MG A529682-250MG
77017-462EA 99 USD
77017-462 77017-464
6-Chloro-3-hydroxybenzo[b]thiophene-2-carboxylic acid 95%
6-Chloro-3-hydroxybenzo[b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅ClO₃S
MDL Number: MFCD25958795
CAS Number: 1393803-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR