4-Chlorobutane-1-sulfonyl chloride 96%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-1-butylsulphonyl Chloride

A529603-250MG A529603-25G A529603-10G
77004-924EA 23 USD
77004-924 77673-404 77882-294
4-Chlorobutane-1-sulfonyl chloride 96%
4-Chlorobutane-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₄H₈Cl₂O₂S
MDL Number: MFCD09952390
CAS Number: 1633-84-7
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR