(2S,3S,4S,5R,6S)-3,4,5-Trihydroxy-6-((5-hydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-7-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid 98+%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Apigenin 7-Glucuronide

A529360-10MG A529360-100MG A529360-50MG
76991-522EA 66.84 USD
76991-522 77684-369 77684-370
(2S,3S,4S,5R,6S)-3,4,5-Trihydroxy-6-((5-hydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-7-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid 98+%, Ambeed.Inc
(2S,3S,4S,5R,6S)-3,4,5-Trihydroxy-6-((5-hydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-7-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid

Precaución: For research use only.

Formula: C₂₁H₁₈O₁₁
MW: 446.36 g/mol
MDL Number: MFCD23699539
CAS Number: 29741-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR