4-Bromo-2-pyridinecarboxamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromopicolinamide
A529207-100MG
A529207-10G
A529207-1G
A529207-250MG
A529207-25G
A529207-5G
77034-730EA
16.98
USD
77034-730
77034-732
77034-734
77034-736
77034-738
77034-740
4-Bromo-2-pyridinecarboxamide 98%
4-Bromo-2-pyridinecarboxamide
Precaución: for Research Use Only
Formula:
C₆H₅BrN₂O MW: 201.03 g/mol |
MDL Number:
MFCD04113813 CAS Number: 62150-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...