5-(2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate hydrochloride 99%

Supplier: Ambeed
Warning

Synonyms: Prasugrel HCl

A529076-100MG A529076-10MG A529076-1MG A529076-250MG A529076-25MG A529076-50MG
77034-718EA 37 USD
77034-718 77034-720 77034-722 77034-724 77034-726 77034-728
5-(2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate hydrochloride 99%
5-(2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₁ClFNO₃S
MDL Number: MFCD11867705
CAS Number: 389574-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR