1-(2,5-Dimethoxyphenyl)ethanol 96%

Supplier: Ambeed
Warning

A529071-10G A529071-5G
77004-914EA 167.63 USD
77004-914 77004-916
1-(2,5-Dimethoxyphenyl)ethanol 96%
1-(2,5-Dimethoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MW: 182.216 g/mol
MDL Number: MFCD08442838
CAS Number: 41038-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR