5-Methylfuran-3(2H)-one 95%

Supplier: Ambeed
Danger

A528649-100MG A528649-250MG
77017-458EA 39 USD
77017-458 77017-460
5-Methylfuran-3(2H)-one 95%
5-Methylfuran-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₆O₂
MDL Number: MFCD15143719
CAS Number: 3511-32-8
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR