4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

A527960-100MG A527960-1G A527960-250MG
77017-446EA 21 USD
77017-446 77017-448 77017-450
4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester 98%
4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₄BNO₄
MW: 305.18 g/mol
MDL Number: MFCD09266188
CAS Number: 1073353-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR