4-(2-Chloro-6-methylpyrimidin-4-yl)morpholine 95%

Supplier: Ambeed
Warning

A527905-100MG A527905-1G A527905-250MG
77034-710EA 67.9 USD
77034-710 77034-712 77034-714
4-(2-Chloro-6-methylpyrimidin-4-yl)morpholine 95%
4-(2-Chloro-6-methylpyrimidin-4-yl)morpholine

Precaución: for Research Use Only

Formula: C₉H₁₂ClN₃O
MW: 213.66 g/mol
MDL Number: MFCD04273703
CAS Number: 52026-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR