1,1-Bis(4-aminophenyl)cyclohexane 98%

Supplier: Ambeed
Warning

A527853-1G A527853-25G A527853-5G A527853-100G
77034-700EA 20 USD
77034-700 77034-702 77034-704 77872-890
1,1-Bis(4-aminophenyl)cyclohexane 98%
1,1-Bis(4-aminophenyl)cyclohexane

Precaución: for Research Use Only

Formula: C₁₈H₂₂N₂
MDL Number: MFCD00035700
CAS Number: 3282-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR