2-[[2-(Dimethylamino)ethyl]methylamino]ethanol 98%

Supplier: Ambeed
Warning

Synonyms: N'-(2-Hydroxyethyl)-N,N,N'-trimethylethylenediamine, N2-(2-Hydroxyethyl)-N1,N1,N2-trimethyl-1,2-ethylenediamine

A527631-100G A527631-25G
77004-912EA 22 USD
77004-912 77882-287
2-[[2-(Dimethylamino)ethyl]methylamino]ethanol 98%
2-[[2-(Dimethylamino)ethyl]methylamino]ethanol

Precaución: for Research Use Only

Formula: C₇H₁₈N₂O
MW: 146.23 g/mol
MDL Number: MFCD00010387
CAS Number: 2212-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR