2-Methoxyethylsulfonyl chloride 97%

Supplier: Ambeed
Danger

A527539-5G A527539-100MG A527539-1G
77004-910EA 213 USD
77004-910 77882-285 77882-286
2-Methoxyethylsulfonyl chloride 97%
2-Methoxyethylsulfonyl chloride

Precaución: for Research Use Only

Formula: C₃H₇ClO₃S
MDL Number: MFCD10032391
CAS Number: 51517-01-2
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR