(R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone 98%

Supplier: Ambeed
Warning

A527468-100MG A527468-1G A527468-250MG A527468-5G
77004-904EA 14 USD
77004-904 77004-906 77004-908 77594-632
(R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone 98%
(R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₂
MDL Number: MFCD03093555
CAS Number: 191090-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR