Methyl 2-(5-amino-2-methylphenyl)acetate 97%

Supplier: Ambeed
Warning

A527304-100MG A527304-250MG
77034-694EA 46.68 USD
77034-694 77034-696
Methyl 2-(5-amino-2-methylphenyl)acetate 97%
Methyl 2-(5-amino-2-methylphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.21 g/mol
MDL Number: MFCD11655592
CAS Number: 850449-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR