3-Methoxyphenethyl bromide 97%

Supplier: Ambeed
Danger

A526906-10G A526906-25G A526906-5G A526906-1G
77004-898EA 400 USD
77004-898 77004-900 77004-902 77882-284
3-Methoxyphenethyl bromide 97%
3-Methoxyphenethyl bromide

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MDL Number: MFCD07369789
CAS Number: 2146-61-4
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR