5-Methoxyquinazoline-2,4-diamine 95%

Supplier: Ambeed
Warning

Synonyms: 2,4-Diamino-5-methoxyquinazoline, 2,4-Diamino-5-methoxy-1,3-benzodiazine

A526611-1G A526611-250MG
77017-434EA 204.76 USD
77017-434 77017-436
5-Methoxyquinazoline-2,4-diamine 95%
5-Methoxyquinazoline-2,4-diamine

Precaución: for Research Use Only

Formula: C₉H₁₀N₄O
MW: 190.202 g/mol
MDL Number: MFCD00665830
CAS Number: 27018-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR