Stearyl gallate 98%

Supplier: Ambeed
Warning

Synonyms: Gallic Acid Stearyl Ester, Octadecyl 3,4,5-trihydroxybenzoate

A526445-100G A526445-25G A526445-5G A526445-500G
77034-688EA 94 USD
77034-688 77034-690 77034-692 77882-283
Stearyl gallate 98%
Stearyl gallate

Precaución: for Research Use Only

Formula: C₂₅H₄₂O₅
MW: 422.61 g/mol
MDL Number: MFCD00016429
CAS Number: 10361-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR