7-Fluoroindoline hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A526399-100MG
A526399-1G
A526399-250MG
77017-428EA
40
USD
77017-428
77017-430
77017-432
7-Fluoroindoline hydrochloride 97%
7-Fluoroindoline hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₈FN·ClH |
MDL Number:
MFCD27923560 CAS Number: 1820711-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...