(4-(Bromomethyl)phenoxy)(tert-butyl)dimethylsilane 95%

Supplier: Ambeed
Danger

A526111-1G A526111-5G
76991-502EA 143.23 USD
76991-502 76991-504
(4-(Bromomethyl)phenoxy)(tert-butyl)dimethylsilane 95%
(4-(Bromomethyl)phenoxy)(tert-butyl)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₃H₂₁BrOSi
MW: 301.29 g/mol
MDL Number: MFCD27920682
CAS Number: 120506-39-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR