Octadecylamine 90%

Supplier: Ambeed
Warning

Synonyms: Octadecan-1-amine, 1-Octadecylamine, Stearylamine, 1-Aminooctadecane

A525938-1000G A525938-100G A525938-25G A525938-500G
77004-886EA 35 USD
77004-886 77004-888 77004-890 77004-892
Octadecylamine 90%
Octadecylamine

Precaución: for Research Use Only

Formula: H₃C(CH₂)₁₆CH₂NH₂
MW: 269.51 g/mol
Boiling Pt: 349
Flash Pt: 110
MDL Number: MFCD00008159
CAS Number: 124-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR