1-Boc-4-ethylaminopiperidine 95%

Supplier: Ambeed
Danger

Synonyms: 1-Boc-4-Ethylaminopiperidine, 4-(Ethylamino)piperidine, N1-BOC protected

A525598-1G A525598-250MG A525598-5G
77004-882EA 30 USD
77004-882 77004-884 77882-281
1-Boc-4-ethylaminopiperidine 95%
1-Boc-4-ethylaminopiperidine

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD06808580
CAS Number: 264905-39-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR