5-(2-Chlorophenyl)-3-isoxazolecarboxylic acid 98+%

Supplier: Ambeed
Warning

A525498-100MG A525498-1G A525498-250MG A525498-5G
77004-874EA 8 USD
77004-874 77004-876 77004-878 77004-880
5-(2-Chlorophenyl)-3-isoxazolecarboxylic acid 98+%
5-(2-Chlorophenyl)-3-isoxazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₃
MDL Number: MFCD06005852
CAS Number: 334017-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR