Fmoc-Trp(For)-OH 98%

Supplier: Ambeed
Warning

A525485-1G A525485-5G
77004-870EA 45.62 USD
77004-870 77004-872
Fmoc-Trp(For)-OH 98%
Fmoc-Trp(For)-OH

Precaución: for Research Use Only

Formula: C₂₇H₂₂N₂O₅
MDL Number: MFCD08458583
CAS Number: 152338-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR