3,5-Difluoro-2-hydroxybenzonitrile 95%
Supplier: Ambeed
|
Danger
|
A524877-100MG
A524877-1G
A524877-250MG
76991-486EA
26
USD
76991-486
76991-488
76991-490
3,5-Difluoro-2-hydroxybenzonitrile 95%
3,5-Difluoro-2-hydroxybenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃F₂NO |
MDL Number:
MFCD08275288 CAS Number: 862088-17-3 UN: 3439 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...