(2-Nitrobenzyl)triphenylphosphonium bromide monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: (2-Nitrobenzyl)triphenylphosphonium bromide

A524479-1G A524479-250MG A524479-25G A524479-5G
77034-672EA 28.65 USD
77034-672 77034-674 77034-676 77034-678
(2-Nitrobenzyl)triphenylphosphonium bromide monohydrate 98%
(2-Nitrobenzyl)triphenylphosphonium bromide monohydrate

Precaución: for Research Use Only

Formula: C₂₅H₂₁BrNO₂P
MW: 478.326 g/mol
MDL Number: MFCD04971966
CAS Number: 23308-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR